Order Entry
Export
ContactUsLinkComponent
L(+)-Lysine dihydrochloride, Sigma-Aldrich®
L(+)-Lysine dihydrochloride, Sigma-Aldrich®
Catalog # SIALL5751-100G
Supplier:  Merck
CAS Number:  
L(+)-Lysine dihydrochloride, Sigma-Aldrich®
Catalog # SIALL5751-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :219.11 g/mol
Melting Pt:200.2 °C
 :M-Clarity™ quality level = MQ200
EINECS:211-518-3
CAS Number:657-26-1
Formula:C₆H₁₆Cl₂N₂O₂
MDL Number:MFCD00066389