Order Entry
Export
ContactUsLinkComponent
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Catalog # MOLE12705461-100G
Supplier:  Molekula
CAS Number:  
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Catalog # MOLE12705461-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :213.25 g/mol
Merck Index:13,05929
Melting Pt:> 300 °C
Storage Temperature:Ambient
EINECS:224-632-3
CAS Number:145224-94-8
Formula:C₆H₁₃NO₄S·1H₂O
MDL Number:MFCD00149409