Order Entry
Export
ContactUsLinkComponent
Sodium diethyldithiocarbamate trihydrate
Sodium diethyldithiocarbamate trihydrate
Catalog # EHERC12605750
Supplier:  EHRENSTORFER
CAS Number:  
Sodium diethyldithiocarbamate trihydrate
Catalog # EHERC12605750
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :225.31 g/mol
Merck Index:12,03443
Melting Pt:95…98.5 °C
Density:1.1 g/cm³ (20 °C)
EINECS:205-710-6
UN:3077
ADR:9,III
 :GHS07
 :GHS09
CAS Number:20624-25-3
Formula:(C₂H₅)₂NCSSNa·3H₂O
MDL Number:MFCD00150617