Manganese(II) acetate tetrahydrate, Sigma-Aldrich®
Catalog # SIAL229776-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:245.09 g/mol
Merck Index:13,05746
Melting Pt:300 °C
Density:1.58 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:211-334-3
CAS Number:6156-78-1
Formula:(H₃CCOO)₂Mn·4H₂O
MDL Number:MFCD00062552