Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Calcium acetate xhydrate
Calcium acetate xhydrate, Sigma-Aldrich®
Calcium acetate xhydrate, Sigma-Aldrich®
Calcium acetate xhydrate, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Calcium acetate xhydrate, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
114460-21-8
:
5743-26-0
Formula:
(H₃CCOO)₂Ca·nH₂O
MDL Number:
MFCD00150019
:
176.18 g/mol
Density:
1.5 g/cm³ (20 °C)
EINECS:
200-540-9