DL-Cystine
Catalog # MOLE20823164-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Merck Index:13,02811
Boiling Pt:429 °C (1013 hPa)
Melting Pt:227 °C
:240.3 g/mol
EINECS:213-094-5
CAS Number:923-32-0
Formula:C₆H₁₂N₂O₄S₂
MDL Number:MFCD00084652