Order Entry
Export
ContactUsLinkComponent
DL-Cystine
DL-Cystine
Catalog # EHERC11925000
Supplier:  EHRENSTORFER
CAS Number:  
DL-Cystine
Catalog # EHERC11925000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
Merck Index:13,02811
Boiling Pt:429 °C (1013 hPa)
Melting Pt:227 °C
 :240.3 g/mol
EINECS:213-094-5
CAS Number:923-32-0
Formula:C₆H₁₂N₂O₄S₂
MDL Number:MFCD00084652