Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Selenic acid
Selenic acid 40% in aqueous solution, Sigma-Aldrich®
Selenic acid 40% in aqueous solution, Sigma-Aldrich®
Toxic
Health
Environment
Selenic acid 40% in aqueous solution, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Toxic
Health
Environment
Selenic acid 40% in aqueous solution, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
7783-08-6
:
Formula:
H₂O₄Se
MDL Number:
MFCD00012191
:
144.97
:
M-Clarity™ quality level = MQ100
EINECS:
231-979-4
:
GHS06
:
GHS08
:
GHS09