Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Orcein
Orcein analytical reagent
Orcein analytical reagent
Irritant
Orcein analytical reagent
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
Orcein analytical reagent
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
1400-62-0
:
MDL Number:
MFCD00062310
Merck Index:
14,6863
EINECS:
215-750-6
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAB20294
Videos
Thermo Scientific Acroseal 25 Years
Documentation