Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Orcein
Orcein, pure
Orcein, pure
:
Thermo Fisher Scientific
Orcein, pure
:
Thermo Fisher Scientific
:
1400-62-0
:
:
MFCD00062310
:
215-750-6
:
GHS07
:
Ambient
Appearance:
Brown to dark purple to black Powder
Infrared spectrum:
Conforms
Loss on drying:
≤12 %
UV Lambda max:
575 to 583 nm
Absorption Ratio P-15/P+15:
0.98 to 1.18
ACRO41657