Lanolin, Sigma-Aldrich®
Glass bottle for solids, 1 kg
Catalog # SIAL49909-1KG-F
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: Wool fat
- CAS Number:8006-54-0
- MDL Number:MFCD00081740
- Merck Index:12,05371
- Melting Pt:38…44 °C
- Density:0.932…0.945 g/cm³ (15 °C)
- :M-Clarity™ quality level = MQ100
- Flash Pt:113 °C (closed cup)
- EINECS:232-348-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



