Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium silicate
Sodium silicate
Sodium silicate
Irritant
Sodium silicate
Supplier:
Roth Carl
Certificates
Expand image
Irritant
Sodium silicate
Supplier:
Roth Carl
About this item
CAS Number:
1344-09-8
:
Formula:
Na₂O(SiO₂)x·xH₂O
MDL Number:
MFCD00003492
Merck Index:
13,08751
Density:
1.39 g/cm³ (25 °C)
EINECS:
215-687-4
:
GHS07