Order Entry
Export
ContactUsLinkComponent
Bismuth
Bismuth
Catalog # PROONIST-2235
CAS Number:  
Bismuth
Catalog # PROONIST-2235
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1,5 g
  • Seq:
    3092

Specifications

About this item

 :
 :208.98 g/mol
Merck Index:12,01297
Boiling Pt:1490 °C (1013 hPa)
Melting Pt:271…272 °C
Density:9.78 g/cm³ (20 °C)
EINECS:231-177-4
CAS Number:7440-69-9
Formula:Bi
MDL Number:MFCD00134033