

Ethanol denatured 96% ≥96%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:64-17-5
:
Formula:H₃CCH₂OH
MDL Number:MFCD00003568
:46.07 g/mol
Boiling Pt:78 °C (1013 hPa)
Melting Pt:–114 °C
Density:0.807 g/cm³ (25 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
:GHS02
:GHS08
Specification Test Results
- Ethanol:95 - 97 %
- Methanol:3 - 5 %
- Water:5 - 6 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000)
Packaging Information
spec_VWRC9763.1000 spec_VWRC9763.1000 spec_VWRC9763.5000 spec_VWRC9763.5000