Order Entry
Export
ContactUsLinkComponent
 
Silica gel, NORMASIL 60® 40 - 63 µm
  27623.460
 :  
undefined
Silica gel, NORMASIL 60® 40 - 63 µm
  27623.460
 :  

 

  • Pack type:
    Plastic bucket
  • Pk:
    25 kg
  • Seq:
    4283
  • Environmentally Preferable:

 

 

  • pH (20°C; 10 %):
    6.5 - 7.5
  • Loss on drying (150°C):
    Max. 12 %
  • Particle size d10:
    25 to 45 µm
  • Particle size d50:
    50 to 70 µm
  • Particle size d90:
    90 to 115 µm

 

 

 :
 :60.08 g/mol
 :Ambient
 :63231-67-4
 :SiO₂
 :MFCD00011232
 :2230 °C (1013 hPa)
 :> 1600 °C