Pumice stone, powder, TECHNICAL
Plastic bottle, 5 kg
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
- CAS Number:1332-09-8
- MDL Number:MFCD00148147
- Melting Pt:> 500 °C
Documents
Certificates
Search by Lot Number20 L Plastic bucket for solids UN Z (ZPACK13247020)
Packaging Information
28577123.pdf P20_VWRC26396.365_SPEC_EN.pdf spec_VWRC26396.365
Product specification
Specifications
Specification Test Results
- Identification:Passes test
Specifications
- Pack type:Plastic bottle
- Pk:5 kg



