Order Entry
Export
ContactUsLinkComponent
5-Aminoallyl-UTP sodium salt 10 mM in TE buffer
5-Aminoallyl-UTP sodium salt 10 mM in TE buffer
Catalog # 40021.
Supplier:  Biotium
CAS Number:  
5-Aminoallyl-UTP sodium salt 10 mM in TE buffer
Catalog # 40021.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pack type:
    Plastic vial
  • Pk:
    100 µl
  • Seq:
    1286

Specifications

About this item

 :
 :605.17 g/mol
Storage Temperature:Freezer
CAS Number:112131-73-4
Formula:C₁₂H₁₇N₃Na₃O₁₅P₃