Order Entry
Export
ContactUsLinkComponent
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) ≥98%, GPR RECTAPUR®
Irritant
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98%
  • Melting point:
    36 - 40 °C

Specification Test Results