Anti-ORF6 Rabbit Polyclonal Antibody
Catalog # BWRLNCP0013
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:ORF6
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:2019-nCoV
- Gene ID:27202..27387
- Accession no.:QHD43420.1
- Storage buffer:Rabbit IgG, 1mg/ml in PBS with 0.02% sodium azide, pH7.2
- Storage temperature:Store at 4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles.
- Immunogen:ORF6 protein, synthetic
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ORF6 polyclonal antibody detects endogenous levels of ORF6 protein
- For research and diagnosis
Type: Primary
Antigen: ORF6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: 2019-nCoV