Order Entry
Export
ContactUsLinkComponent
Barium acetate 99.0-102.0%, AnalaR NORMAPUR® ACS for analysis
Irritant
Barium acetate 99.0-102.0%, AnalaR NORMAPUR® ACS for analysis
Catalog # 85980.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Barium acetate 99.0-102.0%, AnalaR NORMAPUR® ACS for analysis
Catalog # 85980.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    99,0 - 102,0 %
  • pH (5 %) :
    7,0 - 8,5
  • Heavy metals (as Pb) :
    Max. 5 ppm
  • Insoluble matter :
    Max. 100 ppm
  • Oxidising substances (as NO₃) :
    Max. 50 ppm
  • Substances not precipitated by H₂SO₄:
    Max. 0,1 %
  • Cl (Chloride) :
    Max. 10 ppm
  • Ca (Calcium) :
    Max. 0,05 %
  • Fe (Iron) :
    Max. 5 ppm
  • K (Potassium) :
    Max. 30 ppm
  • Na (Sodium) :
    Max. 50 ppm
  • Sr (Strontium) :
    Max. 0,2 %

Specification Test Results

About this item

 :
 :255.42 g/mol
Storage Temperature:Ambient
UN:1564
ADR:6.1,III
 :GHS07
CAS Number:543-80-6
Formula:(H₃CCOO)₂Ba
MDL Number:MFCD00012447