
Isoamyl acetate ≥98%, GPR RECTAPUR®
:123-92-2
:
:C₇H₁₄O₂
:MFCD00008946
:130.19 g/mol
:12,05125
:142 °C (1013 hPa)
:–78 °C
:0.87 g/cm³ (20 °C)
:25 °C
:Ambient
:204-662-3
:1104
:3,III
:GHS02
- Assay:Min. 98 %
- Density (20/4):0.872 --> 0.873
- Free acid (as CH3COOH):Max. 0.1 %
- Isoamyl alcohol:Max. 1.0 %
- Water:Max. 0.1 %