- For:4× E4™ XLS(+) pipettes
- Description:SmartStand SCS-B
- Environmentally Preferable:
- Pk:1
The SmartStand SCS-B is a charging pipette stand for 4 RAININ RFID enabled pipettes, both manual and electronic.
- Unique asset management capability with EasyDirect™ Pipette Asset Management software
- Connects to PC via built-in Bluetooth® wireless technology
- Configurable as stand or hang-up
- Shows service status
Charges 4 E4™ XLS(+) pipettes (110-240 VAC, 50/60 Hz, 0,5 A). Reads and writes the RFID tag of 4 pipettes.