


Mercury(II) thiocyanate ≥99%, AnalaR NORMAPUR® Reag. Ph. Eur.
Catalog # 85965.130
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Assay:Min. 99 %
- Fe (Iron):Max. 20 ppm
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
About this item
:
Merck Index:13,05917
Melting Pt:–38.9 °C
Density:4 g/cm³ (20 °C)
Flash Pt:120 °C
Storage Temperature:Ambient
EINECS:209-773-0
UN:1646
ADR:6.1,II
:316.76 g/mol
:GHS06
:GHS08
:GHS09
CAS Number:592-85-8
Formula:C₂HgN₂S₂
MDL Number:MFCD00011039