Order Entry
Export
ContactUsLinkComponent
DNA/RNA oxidative damage (high sensitivity) ELISA kit
DNA/RNA oxidative damage (high sensitivity) ELISA kit
Supplier:  Cayman Chemical
Certificates
undefined
DNA/RNA oxidative damage (high sensitivity) ELISA kit
Supplier:  Cayman Chemical

Specifications

  • Assay Type:
    Competitive
  • Target protein:
    8-hydroxy-2'-Deoxyguanosine (8-OH-DG)
  • Cross reactivity:
    (8-Hydroxy-2’-deoxyguanosine) - 100%, (8-hydroxyguanosine) - 38%, (8-hydroxyguanine) - 23%, (Guanosine) - <0.01%
  • Storage temperature:
    –20 °C
  • Sensitivity:
    0.03 ng/ml

Specifications