Nucleic acid purification kit, High Pure Viral Nucleic Acid Kit
Catalog # ROCH11858874001
Supplier: ROCHE
CAS Number:
Specifications
- Pk:1 KIT
- Description:High Pure Viral Nucleic Acid Kit
- Sample size:Up to 700 μL (typical 200 - 600 μL)
Specifications
About this item
The High Pure Viral Nucleic Acid Kit is intended for general laboratory use and efficiently purifies viral nucleic acids from serum, plasma, or whole blood using sample volumes of up to 200 μl. When using whole blood as starting material, total nucleic acids are isolated, including viral nucleic acids. The purified viral nucleic acid is eluted in nuclease-free water, and is suitable for direct use in PCR and RT-PCR.
- Isolate both RNA and DNA, allowing simultaneous detection of both types of virus
- Remove inhibitors that might interfere with downstream assays, ensuring greater assay specificity, sensitivity, and reproducibility
- Obtain a concentrated sample, suitable for direct assay (no precipitation required)
- Scale the procedure to accommodate samples with low viral loads
- Save time with a kit that prepares multiple PCR/RT-PCR templates in just minutes
- Eliminate the use of hazardous organic compounds such as cesium chloride, phenol, chloroform, and ethidium bromide
Delivery: Kit contains the following components: Binding Buffer; Poly (A), lyophilised; Proteinase K, recombinant, lyophilisate; Inhibitor Removal Buffer; Wash Buffer; Elution Buffer; High Pure Spin Filter Tubes (containing glass fiber fleece); Collection Tubes.