Avantor® Hichrom, Tubing Cutters, Stainless Steel
Catalog # HICHHI-192
Supplier: Avantor
Specifications
- Description:Stainless steel tubing cutter
- Seq:01
- Pk:1
Specifications
About this item
Stainless steel cutters are specifically designed for ¹/₁₆" stainless steel tubing.
- Smooth and uniform cuts