Order Entry
Export
ContactUsLinkComponent
Lucifer yellow CH dilithium salt 100 mM in water fluorescent dye
Lucifer yellow CH dilithium salt 100 mM in water fluorescent dye
Catalog # BTIU80026
Supplier:  Biotium
CAS Number:  
undefined
Lucifer yellow CH dilithium salt 100 mM in water fluorescent dye
Catalog # BTIU80026
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    100 µl
  • Seq:
    1286

Specifications

About this item

 :
 :457.25 g/mol
Storage Temperature:Ambient
CAS Number:67769-47-5
Formula:C₁₃H₉N₅O₉S₂Li₂
MDL Number:MFCD00006922