Order Entry
Export
ContactUsLinkComponent
Chinese ink, green, Q Path® stain for histology
Chinese ink, green, Q Path® stain for histology
Catalog # 01816201.
Supplier:  VWR Chemicals
CAS Number:  
undefined
Chinese ink, green, Q Path® stain for histology
Catalog # 01816201.
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    30 mL
  • Seq:
    2345

Specifications

Specification Test Results

  • Appearance:
    Green liquid
  • Colour of solution:
    Passes test
  • Density (20/4):
    1.190 --> 1.370

Specification Test Results

About this item

 :
 :1333-86-4