Order Entry
Export
ContactUsLinkComponent
8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt fluorescent dye
Irritant
8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt fluorescent dye
Catalog # 90028.
Supplier:  Biotium
CAS Number:  
undefined
Irritant
8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt fluorescent dye
Catalog # 90028.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :523.40 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:196504-57-1
Formula:C₁₆H₈NNa₃O₉S₃
MDL Number:MFCD00674494