Order Entry
Export
ContactUsLinkComponent
 
RH 414 membrane stain, for histology
Catalog # 61016.
Supplier:  Biotium
CAS Number:  
undefined
RH 414 membrane stain, for histology
Catalog # 61016.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :581.476 g/mol
Storage Temperature:Freezer
CAS Number:161433-30-3
Formula:C₂₈H₄₃Br₂N₃