Di-8-ANEPPS membrane stain, for histology
Catalog # 61012.
Supplier: Biotium
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:592.87 g/mol
Storage Temperature:Refrigerator
CAS Number:157134-53-7
Formula:C₃₆H₅₂N₂O₃S
MDL Number:MFCD03453608