Phloridzin ≥98%, crystalline solid
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:60-81-1
:
Formula:C21H24O10
MDL Number:MFCD00006591
:436.41 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity:≥98%
- Formulation:A Crystalline Solid
- λmax:224, 285 nm