ContactUsLinkComponent
Phthalate-phosphate reagent powder pillows
Phthalate-phosphate reagent powder pillows
Catalog # HACH2150166
Supplier:  Hach
CAS Number:  
undefined
Phthalate-phosphate reagent powder pillows
Catalog # HACH2150166
Supplier:  Hach
CAS Number:  
Specifications
  • Chemical method:
    PAN
  • Pk:
    50
  • Description:
    Phthalate-phosphate reagent powder pillows
  • Sample size:
    25 ml
Specifications
About this item
For low-level determination of nickel and cobalt by the PAN method.
Packaging: Pack of 50 powder pillows.