Order Entry
Export
Orders LinkContactUsLinkComponent
Qualitative Test Paper, Watesmo

Qualitative Test Paper, Watesmo

Udder test paper for mastitis, 0, Color reaction with a pH indicator, 100×140×7 mm (packaging), 20 Tests, 20
Catalog # MANA90748
Supplier:   MACHEREY-NAGEL
Qualitative Test Paper, Watesmo
Catalog # MANA90748
Supplier:  MACHEREY-NAGEL
CAS Number:  
Qualitative Test Paper, Watesmo
Qualitative Test Paper, Watesmo

Udder test paper for mastitis, 0, Color reaction with a pH indicator, 100×140×7 mm (packaging), 20 Tests, 20

CAS Number:  
Price
8,25 €
Log in for availability
1 * 20 items
Product Details & Documents

The test paper allows the quick and easy detection of water and water vapor.

  • Colour change: Light blue to dark blue

On contact with water the paper changes colour from light blue to dark blue. The reaction is not reversible. Even after drying the contact with water can clearly be proved.

Specifications

Specifications

  • Chemical method:
    Color reaction with a pH indicator
  • Pk:
    20
  • Description:
    Udder test paper for mastitis
  • Dimensions:
    100×140×7 mm (packaging)
  • Tests per kit:
    20 Tests
  • Range:
    0
  • Cat. No.:
    MANA90748
  • Sensitivity:
    Mastitis - Traces
  • Contents:
    20 sheets in PE bag
  • Platform:
    Qualitative test paper
  • Compliance:
    Not CE certified and no hazardous material
  • Shelf life (months):
    24 (from production)
  • W×D×H:
    90×140 mm (net)
  • Weight:
    48,13 g

Specifications