Test strips, qualitative, nickel
Catalog # MANA90730
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Chemical method:Diacetyldioxime
- Pk:200
- Description:Nickel (II) ions
Specifications
About this item
This test paper allows the quick and easy detection of Nickel.
- Colour changes: White to red
For non-destructive testing of materials apply a drop of 3% Nitric acid (1 part Nitric acid 85%, 5 parts water) to the degreased surface. After 1 minute absorb the liquid with the test paper. With this procedure Nickel contents >0,5 % are safely detected.
Limit of sensitivity: 10 mg/L Ni2+
Packaging: Pack of 200 strips 20×70 mm