Order Entry
Export
ContactUsLinkComponent
Cresyl violet (acetate), certified by the Biological Stain Commission stain for cytology, Sigma-Aldrich®
Cresyl violet (acetate), certified by the Biological Stain Commission stain for cytology, Sigma-Aldrich®
Catalog # C5042-10G
CAS Number:  
undefined
Cresyl violet (acetate), certified by the Biological Stain Commission stain for cytology, Sigma-Aldrich®
Catalog # C5042-10G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :321.33 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
CAS Number:10510-54-0
Formula:C₁₈H₁₅N₃O₃
MDL Number:MFCD00013151