ContactUsLinkComponent
VISOCOLOR® Colorimetric Test, pH
undefined
VISOCOLOR® Colorimetric Test, pH
  • Model:
    MANA920174
  • Platform:
    Colorimetric test kits
  • Storage temperature:
    15…25 °C
  • Shelf life (months):
    24
  • Weight:
    142 g (including packaging)