5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE) ≥80% (by HPLC), BioReagent suitable for fluorescence, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:117548-22-8
:76608-16-7
Formula:C₂₅H₁₅NO₉
MDL Number:MFCD04119713
:473.39 g/mol