Serotonin ELISA kit
Catalog # ABNOKA2518
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Target protein:Serotonin
- Description:Serotonin ELISA kit
- Storage conditions:4 °C
Specifications
About this item
Serotonin ELISA kit is an ultra-sensitive enzyme immunoassay for the quantitative determination of serotonin.
- Calibration range: 015 - 2.5 ng/ml
- Sample volume: 100 µl
- Detection method: colorimetric
- Label: HRP-conjugate
Delivery: Supplied with acylation plate (96 wells), stabilizer10% solution (4 ml), dilution concentrate 20X (50 ml), wash buffer concentrate 50X (20 ml), enzyme conjugate (12 ml), substrate (12 ml), stop solution (12 ml), adhesive foil (4 slices), serotonin antiserum (3 ml), acylation buffer (4 ml), serotonin microtiter strips 96(8×12)wells.
Caution: For research use only (RUO).