Orders LinkContactUsLinkComponent
LED excitation modules
LED excitation modules
Supplier:   MOTIC
LED excitation modules
Supplier:  MOTIC
Certificates
LED excitation modules
LED excitation modules
3 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Availability
    Price
  • MOTI1101011400251
    LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube
    LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 700,00 €
    1 700,00 €
    1 * 1 items
       1 700,00 €
  • MOTI1101011400241
    LED excitation module FITC, consisting of 470 nm LED lamp and FITC filter cube
    LED excitation module FITC, consisting of 470 nm LED lamp and FITC filter cube
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 480,00 €
    1 480,00 €
    1 * 1 items
       1 480,00 €
  • MOTI1101011400231
    LED excitation module TRITC, consisting of 525 nm LED lamp and TRITC filter cube
    LED excitation module TRITC, consisting of 525 nm LED lamp and TRITC filter cube
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 480,00 €
    1 480,00 €
    1 * 1 items
       1 480,00 €