ESD safety shoes, lace-up, Matthew Pro GTX Low
Supplier: ELTEN
Certificates
About this item
Leather-free ESD safety shoes made from hydrophobised microfibre and hydrophobised textile material CORDURA® leather with GORE-TEX performance, a fully padded bellows tongue and a dual densitiy PU/PU sole ERGO-ACTIVE 2.
- Climate-regulating, breathable textile lining
- Replaceable insole; orthopaedic insoles possible
- Semi-orthopaedic inlay sole ESD black
- Anatomically-shaped insole and ankle support for improved stability
- TPU scuff cap
- Metal-free, puncture resistant midsole
- Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects. The sizes
36 - 39 are ideally tailored to the ergonomics of female feet.
Colour: Black/red
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2 is suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3 is suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certifications: DGUV 112-191; EN 61340-5-1 (ESD); EN ISO 20345 S3 CI/SRC, Form A
Specifications
- Catalog No:
- ELTN7278101/47
- ELTN7278101/46
- ELTN7278101/45
- ELTN7278101/44
- ELTN7278103/48
- ELTN7278101/43
- ELTN7278103/47
- ELTN7278103/46
- ELTN7278103/45
- ELTN7278102/43
- ELTN7278101/41
- ELTN7278102/42
- ELTN7278101/40
- ELTN7278102/41
- ELTN7278102/40
- ELTN7278101/48
- ELTN1727803/39
- ELTN1727802/38
- ELTN1727803/37
- ELTN1727803/38
- ELTN1727802/39
- ELTN1727803/36
- ELTN7278103/40
- ELTN7278102/48
- ELTN7278102/47
- ELTN7278102/46
- ELTN7278102/45
- ELTN7278102/44
- ELTN7278101/42
- ELTN7278103/44
- ELTN7278103/43
- ELTN7278103/42
- ELTN7278103/41
- ELTN1727801/38
- ELTN1727801/39
- ELTN1727801/36
- ELTN1727801/37
- ELTN1727802/36
- ELTN1727802/37
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001