ESD safety shoes, lace-up, Mason Pro GTX Mid
: ELTEN
Lightweight and comfortable ESD safety ankle boots made from cowhide leather, GORE-TEX® lining, reflective materials, a full padded bellows tongue and a dual density PU-PU sole ERGO-ACTIVE 2.
- GORE-TEX® climate-regulating, breathable textile lining, water resistance
- Semi-orthopaedic inlay sole ESD black
- Metal-free, puncture resistant midsole
- TPU scuff cap
- Protective steel toe cap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/Blue
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
: EN 61340-5-1(ESD); DGUV 112-191 (BGR 191); EN ISO 20345; S3 SRC, form B
- Catalog No:
- ELTN7683102/43
- ELTN7683102/42
- ELTN7683102/41
- ELTN7683102/40
- ELTN7683101/48
- ELTN7683101/47
- ELTN7683101/46
- ELTN7683102/46
- ELTN7683102/45
- ELTN7683102/44
- ELTN7683101/41
- ELTN7683103/45
- ELTN7683101/40
- ELTN7683103/44
- ELTN7683103/43
- ELTN7683103/42
- ELTN7683103/41
- ELTN7683103/40
- ELTN7683102/48
- ELTN7683102/47
- ELTN7683101/45
- ELTN7683101/44
- ELTN7683101/43
- ELTN7683103/47
- ELTN7683101/42
- ELTN7683103/46
- ELTN7683103/48
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001