Order Entry
Export
ContactUsLinkComponent
(±)-Geosmin solution 2 mg/ml in methanol ≥97% (by GC), Sigma-Aldrich®
Flammable
Toxic
Health
(±)-Geosmin solution 2 mg/ml in methanol ≥97% (by GC), Sigma-Aldrich®
Catalog # SIALG5908-1ML
Supplier:  Merck
CAS Number:  
Flammable
Toxic
Health
(±)-Geosmin solution 2 mg/ml in methanol ≥97% (by GC), Sigma-Aldrich®
Catalog # SIALG5908-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

Specification Test Results

  • Assay (Calulated on Dry Substance, GC):
    ≥97%

Specification Test Results

About this item

 :
 :182.30 g/mol
Flash Pt:11 °C
Storage Temperature:Freezer
EINECS:200-659-6
UN:1230
ADR:3,II
 :GHS02
 :GHS06
 :GHS08
CAS Number:16423-19-1
Formula:C₁₂H₂₂O
MDL Number:MFCD00055578