Order Entry
Export
Orders LinkContactUsLinkComponent
QUANTOFIX® Test Strips, Ag-Fix

QUANTOFIX® Test Strips, Ag-Fix

QUANTOFIX® active oxygen, 0 • 4 • 8 • 15 • 25 - mg/L - KMPS, Redox reaction (potassium iodide/redox indicator), 47×126×40 mm (packaging), 100 Tests, 100
Catalog # MANA91349
Supplier:   MACHEREY-NAGEL
QUANTOFIX® Test Strips, Ag-Fix
Catalog # MANA91349
Supplier:  MACHEREY-NAGEL
CAS Number:  
QUANTOFIX® Test Strips, Ag-Fix
QUANTOFIX® Test Strips, Ag-Fix

QUANTOFIX® active oxygen, 0 • 4 • 8 • 15 • 25 - mg/L - KMPS, Redox reaction (potassium iodide/redox indicator), 47×126×40 mm (packaging), 100 Tests, 100

CAS Number:  
Price
39,90 €
Log in for availability
1 * 100 items
Product Details & Documents

With these test strips two paper zones are sealed onto the lower end of a plastic strip, one for the determination of the silver concentration and one for pH measurement. After dipping the test strip in the test solution the colours of the test zones are compared with the colour scale on the tube.

  • Always ready to use: no accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity The length of the strips has been chosen for safe and convenient handling

During operation of a fixing bath it is necessary to check the silver content as well as the pH in regular intervals to insure proper operation. A decrease in fixing capacity is noted as an increase in Silver concentration or an alteration of the pH value.

Specifications

Specifications

  • Chemical method:
    Redox reaction (potassium iodide/redox indicator)
  • Pk:
    100
  • Description:
    QUANTOFIX® active oxygen
  • Dimensions:
    47×126×40 mm (packaging)
  • Tests per kit:
    100 Tests
  • Range:
    0 • 4 • 8 • 15 • 25 - mg/L - KMPS
  • Cat. No.:
    MANA91349
  • Parameters:
    All reagents included
  • Contents:
    100 test strips in a tube, instruction leaflet
  • Platform:
    Semi-quantitative test strips
  • Compliance:
    No hazardous material
  • Instrument:
    No
  • Shelf life (months):
    30 (from production)
  • Method:
    Active oxygen - Redox reaction (yellow → green)
  • Weight:
    45 g

Specifications