Ultipor® GF Plus Filter Cartridge
Supplier: Cytiva (Formerly Pall Lab)
Certificates
About this item
Pall Ultipor® GF Plus filter cartridges are made with high voidage glass fiber media for enhanced filter life. The cartridges feature a high area pleated element construction designed to fit in sanitary-style filter housings.
- High area pleated medium
- Low differential pressures
- Absolute particle-rated in liquids
- High-capacity for long-life
- Fixed pores prevent unloading
- Choice of AB (SOE) sanitary or (DOE) cartridge styles
- Manufactured for use in conformance with cGMP
A bio-safe low extractables binder resin covalently immobilises the glass fibers and imparts high-strength and reliable removal efficiency. Due to the high particulate removal efficiency and low pressure drops consistent with glass fiber media, Ultipor GF Plus filters effectively protect sterilising-grade filters in air and gas applications.
Certifications: Ultipor GF Plus filters are manufactured in accordance with ISO 9000 and GMP requirements. All components meet the specifications for biological tests listed in the USP for Class VI plastics at 121 °C. Each box contains a certificate which confirms the quality standards in manufacturing, the traceability of all components, and the above mentioned safety tests. Each filter is supplied with a certificate of test that confirms suitability for use in pharmaceutical applications. Samples are tested for effluent quality to meet the USP requirements regarding Total Organic Carbon, pH shift and Endotoxins.
Specifications
- Catalog No:
- PALLU010Z14225
- 516-9822
- Max. pressure:
- −
- 4 bar
- Max. temperature:
- −
- 140 °C