ContactUsLinkComponent
Industrial gloves, oil repellent, HyFlex® 11-939
Industrial gloves, oil repellent, HyFlex® 11-939
Supplier:  Ansell
Certificates
undefined
Industrial gloves, oil repellent, HyFlex® 11-939
Supplier:  Ansell
Specifications
  • Catalog No:
  • ANSE11-939/11
  • ANSE11-939/6
  • ANSE11-939/7
  • ANSE11-939/8
  • ANSE11-939/10
  • ANSE11-939/9
  • Model:
  • 11-939
  • 11-939
  • 11-939
  • 11-939
  • 11-939
  • 11-939
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Dyneema® Diamond Technology, nylon, Spandex and carbon fibers
  • Dyneema® Diamond Technology, nylon, Spandex and carbon fibers
  • Dyneema® Diamond Technology, nylon, Spandex and carbon fibers
  • Dyneema® Diamond Technology, nylon, Spandex and carbon fibers
  • Dyneema® Diamond Technology, nylon, Spandex and carbon fibers
  • Dyneema® Diamond Technology, nylon, Spandex and carbon fibers
  • Finish:
  • Fully coated
  • Fully coated
  • Fully coated
  • Fully coated
  • Fully coated
  • Fully coated
  • Gauge:
  • 18
  • 18
  • 18
  • 18
  • 18
  • 18
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4X42B)
  • EN 388:2016 (4X42B)
  • EN 388:2016 (4X42B)
  • EN 388:2016 (4X42B)
  • EN 388:2016 (4X42B)
  • EN 388:2016 (4X42B)
  • Anti-static:
  • +
  • +
  • +
  • +
  • +
  • +
Specifications