Order Entry
Export
ContactUsLinkComponent
Succinimidyl ester thiazole orange DNA/RNA binding dye
Succinimidyl ester thiazole orange DNA/RNA binding dye
Catalog # 40073.
Supplier:  Biotium
CAS Number:  
undefined
Succinimidyl ester thiazole orange DNA/RNA binding dye
Catalog # 40073.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :503 g/mol
Storage Temperature:Freezer
Formula:C₂₈H₂₈N₃O₄S