Endo agar
Catalog # SIALE5399-500G
Supplier: Merck
CAS Number:
Specifications
- Pk:500 g
- Description:Endo agar
Specifications
About this item
This agar is selective and differential for Enterobacter, Escherichia coli and Coliforms, Klebsiella, Proteus, Salmonella, Shigella.
- Microbiology grade
- pH: 7,5 ±0,2 (25 °C)
Components are peptic digest of animal tissue 10,00 g/l, lactose 10,00 g/l, dipotassium phosphate 3,50 g/l, sodium sulphite 2,50 g/l, basic Fuchsin 0,50 g/l, agar 15,00 g/l.