
2,5-Dibromopyrazine 95% (by GC)
Catalog # SIAL796603-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance (Colour):White to Yellow and Faint Beige to Beige and Faint Brown to Brown
- Appearance (Form) :Powder or Crystals
- Infrared Spectrum:Conforms to Structure
- Purity (GC):≥94.5%
- Carbon:18.1 - 22.3%
- Nitrogen:10.5 - 13.0%
Specification Test Results
About this item
:
:237.8800 g/mol
:GHS07
CAS Number:23229-26-7
Formula:C₄H₂Br₂N₂
MDL Number:MFCD08061601