13,14-dihydro-15-keto Prostaglandin F₂α ELISA kit
Supplier: Cayman Chemical
Certificates
About this item
This kit is used for the measurement of 13,14-dihydro-15-keto prostaglandin F₂α in plasma.
- Detect 13,14-dihydro-15-keto PGF₂α, a stable metabolite of PGF₂α
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure 13,14-dihydro-15-keto PGF₂α levels down to 16,3 pg/ml
- Reads colorimetric at 405 to 420 nm
PGF₂α is rapidly metabolized to 13,14-dihydro-15-keto PGF₂α in vivo, by the enzymes 15-hydroxy PG dehydrogenase and D13-reductase. Measurement of 13,14-dihydro-15-keto PGF₂α in plasma can be used as a marker of the in vivo production of PGF₂α.
13,14-dihydro-15-keto PGF₂α ELISA is a competitive assay that provides measurements of 13,14-dihydro-15-keto PGF₂α within the range of 2,3 to 5000 pg/ml. The assay has been validated for the measurement of 13,14-dihydro-15-keto PGF₂α in plasma, a common matrix for measurement of this PGF₂α metabolite.
Delivery: Kit contains antiserum, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover, ellman's reagent, tracer and antiserum dye.
Specifications
- Assay Type:Competitive
- Target protein:13,14-dihydro-15-keto Prostaglandin F₂α
- Developmental time:90 - 120 min
- Mid point:120 - 180 pg/ml (50% B/B₀)
- Detection range:2,3 - 5000 pg/ml
- Storage temperature:–20 °C
- Sensitivity:15 pg/ml (80% B/B₀)
- Incubation time:18 hr